Anti-MSRA Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: MSRA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-623
- 77841-624
- Antigen Symbol:
- MSRA
- MSRA
- Antigen Name:
- Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
- Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
- Antigen Synonyms:
- Cytosolic methionine S sulfoxide reductase| Methionine sulphoxide reductase A| Mitochondrial peptide methionine sulfoxide reductase| MSR A| msrA| _HUMAN| peptide met (O) reductase| Peptide Met(O) reductase| Peptide methionine (S) S oxide reductase| Peptide-methionine (S)-S-oxide reductase| PMSR| Protein methionine S oxide reductase| Protein-methionine-S-oxide reductase
- Cytosolic methionine S sulfoxide reductase| Methionine sulphoxide reductase A| Mitochondrial peptide methionine sulfoxide reductase| MSR A| msrA| _HUMAN| peptide met (O) reductase| Peptide Met(O) reductase| Peptide methionine (S) S oxide reductase| Peptide-methionine (S)-S-oxide reductase| PMSR| Protein methionine S oxide reductase| Protein-methionine-S-oxide reductase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJ68
- Q9UJ68
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice