Anti-MBD5 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MBD5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-692
- 77837-691
- Antigen Symbol:
- MBD5
- MBD5
- Antigen Name:
- Methyl-CpG-binding domain protein 5 protein (623-840AA)
- Methyl-CpG-binding domain protein 5 protein (623-840AA)
- Antigen Synonyms:
- 9430004D19Rik|AA536666|AI426407|C030040A15Rik|FLJ11113|FLJ30517|KIAA1461|MBD5|MBD5_HUMAN|methyl CpG binding domain protein 5|Methyl CpG binding protein MBD5|Methyl-CpG-binding domain protein 5|Methyl-CpG-binding protein MBD5|RP23 167N13.1
- 9430004D19Rik|AA536666|AI426407|C030040A15Rik|FLJ11113|FLJ30517|KIAA1461|MBD5|MBD5_HUMAN|methyl CpG binding domain protein 5|Methyl CpG binding protein MBD5|Methyl-CpG-binding domain protein 5|Methyl-CpG-binding protein MBD5|RP23 167N13.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P267
- Q9P267
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice