Anti-PMAIP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PMAIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-549
- 77794-550
- Antigen Symbol:
- PMAIP1
- PMAIP1
- Antigen Name:
- Phorbol-12-myristate-13-acetate-induced protein 1 protein (1-136AA)
- Phorbol-12-myristate-13-acetate-induced protein 1 protein (1-136AA)
- Antigen Synonyms:
- Adult T cell leukemia derived PMA responsive|APR|APR_HUMAN|ATL-derived|Immediate early response protein APR|Immediate-early-response protein APR|NOXA|Phorbol 12 myristate 13 acetate induced protein 1|Phorbol-12-myristate-13-acetate-induced protein 1|PMA induced protein 1|PMA-induced protein 1|PMA-responsive gene|Pmaip1|Protein Noxa
- Adult T cell leukemia derived PMA responsive|APR|APR_HUMAN|ATL-derived|Immediate early response protein APR|Immediate-early-response protein APR|NOXA|Phorbol 12 myristate 13 acetate induced protein 1|Phorbol-12-myristate-13-acetate-induced protein 1|PMA induced protein 1|PMA-induced protein 1|PMA-responsive gene|Pmaip1|Protein Noxa
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13794
- Q13794
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice