Anti-SIAH1 Rabbit Polyclonal Antibody
100 µl, Vial
Catalog # 77799-531
Supplier: AFG Bioscience
Supplier #: A34701-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: SIAH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77799-531
- Reactivity:Human|Mouse|Rat
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:SIAH1
- Gene ID:Q8IUQ4
- Conjugation:Unconjugated
- Antigen Synonyms:E3 ubiquitin-protein ligase SIAH1|FLJ08065|hSIAH1|HUMSIAH|Seven in absentia homolog 1 (Drosophila)|Seven in absentia homolog 1|Siah 1|Siah 1a|Siah E3 ubiquitin protein ligase 1|Siah-1|Siah-1a|Siah1|SIAH1_HUMAN|SIAH1A|Ubiquitin ligase SIAH1
- Antigen Name:Ubiquitin ligase SIAH1
- ELISA:Yes
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



