Anti-SH3GL1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SH3GL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-981
- 77834-982
- Antigen Symbol:
- SH3GL1
- SH3GL1
- Antigen Name:
- Endophilin-A2 protein (219-368AA)
- Endophilin-A2 protein (219-368AA)
- Antigen Synonyms:
- CNSA1|EEN|EEN fusion partner of MLL|Endophilin-2|Endophilin-A2|Extra 11 19 leukemia fusion gene|Extra eleven-nineteen leukemia fusion gene protein|Fusion partner of MLL|MGC111371|SH3 containing Grb 2 like 1 protein|SH3 containing protein EEN|SH3 domain GRB2 like 1|SH3 domain protein 2B|SH3 domain-containing GRB2-like protein 1|SH3D2B|SH3G1_HUMAN|Sh3gl1|SH3P8
- CNSA1|EEN|EEN fusion partner of MLL|Endophilin-2|Endophilin-A2|Extra 11 19 leukemia fusion gene|Extra eleven-nineteen leukemia fusion gene protein|Fusion partner of MLL|MGC111371|SH3 containing Grb 2 like 1 protein|SH3 containing protein EEN|SH3 domain GRB2 like 1|SH3 domain protein 2B|SH3 domain-containing GRB2-like protein 1|SH3D2B|SH3G1_HUMAN|Sh3gl1|SH3P8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99961
- Q99961
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice