Anti-UGT8 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-309
- 77823-308
- Antigen Symbol:
- UGT8
- UGT8
- Antigen Name:
- 2-hydroxyacylsphingosine 1-beta-galactosyltransferase protein (38-402AA)
- 2-hydroxyacylsphingosine 1-beta-galactosyltransferase protein (38-402AA)
- Antigen Synonyms:
- 2 hydroxyacylsphingosine 1 beta galactosyltransferase|2-hydroxyacylsphingosine 1-beta-galactosyltransferase|Ceramide UDP galactosyltransferase|Ceramide UDP-galactosyltransferase|Cerebroside synthase|CGT|CGT_HUMAN|mCerGT|UDP galactose ceramide galactosyltransferase|UDP galactose ceramide galactosyltransferase 8|UDP glycosyltransferase 8|UDP-galactose-ceramide galactosyltransferase|UGT4|Ugt8|Uridine diphosphate glycosyltransferase 8
- 2 hydroxyacylsphingosine 1 beta galactosyltransferase|2-hydroxyacylsphingosine 1-beta-galactosyltransferase|Ceramide UDP galactosyltransferase|Ceramide UDP-galactosyltransferase|Cerebroside synthase|CGT|CGT_HUMAN|mCerGT|UDP galactose ceramide galactosyltransferase|UDP galactose ceramide galactosyltransferase 8|UDP glycosyltransferase 8|UDP-galactose-ceramide galactosyltransferase|UGT4|Ugt8|Uridine diphosphate glycosyltransferase 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16880
- Q16880
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice