Anti-TIGD7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TIGD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-838
- 77830-839
- Antigen Symbol:
- TIGD7
- TIGD7
- Antigen Name:
- Tigger transposable element-derived protein 7 protein (1-300AA)
- Tigger transposable element-derived protein 7 protein (1-300AA)
- Antigen Synonyms:
- jerky homolog like|Sancho|TIGD7|TIGD7_HUMAN|Tigger transposable element-derived protein 7
- jerky homolog like|Sancho|TIGD7|TIGD7_HUMAN|Tigger transposable element-derived protein 7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6NT04
- Q6NT04
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice