Anti-ARL6IP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARL6IP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-595
- 77822-594
- Antigen Symbol:
- ARL6IP1
- ARL6IP1
- Antigen Name:
- ADP-ribosylation factor-like protein 6-interacting protein 1 protein (93-133AA)
- ADP-ribosylation factor-like protein 6-interacting protein 1 protein (93-133AA)
- Antigen Synonyms:
- ADP ribosylation factor like 6 interacting protein|ADP-ribosylation factor-like protein 6-interacting protein 1|Aip-1|Aip1|AR6P1_HUMAN|ARL-6-interacting protein 1|ARL6 interacting protein 1|ARL6IP|ARMER|KIAA0069
- ADP ribosylation factor like 6 interacting protein|ADP-ribosylation factor-like protein 6-interacting protein 1|Aip-1|Aip1|AR6P1_HUMAN|ARL-6-interacting protein 1|ARL6 interacting protein 1|ARL6IP|ARMER|KIAA0069
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15041
- Q15041
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice