Anti-TBCD Rabbit Polyclonal Antibody
Catalog # 77801-943
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Tubulin-specific chaperone D
- Antigen Symbol:TBCD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BTW9
- Antigen Synonyms:Beta tubulin cofactor D|Beta-tubulin cofactor D|KIAA0988|SSD 1|SSD-1|SSD1|tbcd|TBCD_HUMAN|tfcD|Tubulin folding cofactor D|Tubulin specific chaperone D|Tubulin-folding cofactor D|Tubulin-specific chaperone D
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TBCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat