Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-NDUFS5 Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-NDUFS5 Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77805-951
  • 77805-952
  • Antigen Symbol:
  • NDUFS5
  • NDUFS5
  • Antigen Name:
  • NADH dehydrogenase [ubiquinone] iron-sulfur protein 5 protein (2-106AA)
  • NADH dehydrogenase [ubiquinone] iron-sulfur protein 5 protein (2-106AA)
  • Antigen Synonyms:
  • CI 15k|CI-15 kDa|CI15K|Complex I-15 kDa|NADH dehydrogenase (ubiquinone) Fe S protein 5, 15kDa (NADH coenzyme Q reductase)|NADH dehydrogenase [ubiquinone] iron-sulfur protein 5|NADH-ubiquinone oxidoreductase 15 kDa subunit|NADH:ubiquinone oxidoreductase 15 kDa IP subunit|NDUFS5|NDUS5_HUMAN
  • CI 15k|CI-15 kDa|CI15K|Complex I-15 kDa|NADH dehydrogenase (ubiquinone) Fe S protein 5, 15kDa (NADH coenzyme Q reductase)|NADH dehydrogenase [ubiquinone] iron-sulfur protein 5|NADH-ubiquinone oxidoreductase 15 kDa subunit|NADH:ubiquinone oxidoreductase 15 kDa IP subunit|NDUFS5|NDUS5_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O43920
  • O43920
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice