Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP12 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-USP12 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP12 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77804-106
  • 77804-105
  • Antigen Symbol:
  • USP12
  • USP12
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA)
  • Ubiquitin carboxyl-terminal hydrolase 12 protein (1-370AA)
  • Antigen Synonyms:
  • Deubiquitinating enzyme 12|OTTHUMP00000042337|UBH 1|UBH1|Ubiquitin carboxyl terminal hydrolase 12|Ubiquitin carboxyl-terminal hydrolase 12|Ubiquitin hydrolyzing enzyme 1|Ubiquitin specific peptidase 12|Ubiquitin specific processing protease 12|Ubiquitin specific protease 12 like 1|Ubiquitin thioesterase 12|Ubiquitin thiolesterase 12|Ubiquitin-hydrolyzing enzyme 1|Ubiquitin-specific-processing protease 12|UBP12_HUMAN|USP 12|Usp12|USP12L1
  • Deubiquitinating enzyme 12|OTTHUMP00000042337|UBH 1|UBH1|Ubiquitin carboxyl terminal hydrolase 12|Ubiquitin carboxyl-terminal hydrolase 12|Ubiquitin hydrolyzing enzyme 1|Ubiquitin specific peptidase 12|Ubiquitin specific processing protease 12|Ubiquitin specific protease 12 like 1|Ubiquitin thioesterase 12|Ubiquitin thiolesterase 12|Ubiquitin-hydrolyzing enzyme 1|Ubiquitin-specific-processing protease 12|UBP12_HUMAN|USP 12|Usp12|USP12L1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O75317
  • O75317
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together