Anti-RETN Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RETN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77810-605
- 77810-606
- Antigen Symbol:
- RETN
- RETN
- Antigen Name:
- Resistin protein (19-108AA)
- Resistin protein (19-108AA)
- Antigen Synonyms:
- Adipose tissue specific secretory factor|Adipose tissue-specific secretory factor|ADSF|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein|CG5403|Cysteine rich secreted protein A12 alpha like 2|Cysteine rich secreted protein FIZZ3|Cysteine-rich secreted protein A12-alpha-like 2|Cysteine-rich secreted protein FIZZ3|dri|FIZZ 3|FIZZ3|Found in inflammatory zone 3|HXCP 1|HXCP1|MGC126603|MGC126609|PRO1199|Resistin|Resistin delta2|RETN 1|RETN|RETN_HUMAN|RETN1|RSTN|UNQ407|XCP 1|XCP1
- Adipose tissue specific secretory factor|Adipose tissue-specific secretory factor|ADSF|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein|C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein|CG5403|Cysteine rich secreted protein A12 alpha like 2|Cysteine rich secreted protein FIZZ3|Cysteine-rich secreted protein A12-alpha-like 2|Cysteine-rich secreted protein FIZZ3|dri|FIZZ 3|FIZZ3|Found in inflammatory zone 3|HXCP 1|HXCP1|MGC126603|MGC126609|PRO1199|Resistin|Resistin delta2|RETN 1|RETN|RETN_HUMAN|RETN1|RSTN|UNQ407|XCP 1|XCP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HD89
- Q9HD89
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice