Anti-UMPS Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: UMPS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-729
- 77828-730
- Antigen Symbol:
- UMPS
- UMPS
- Antigen Name:
- Uridine 5\\\'-monophosphate synthase protein (314-444AA)
- Uridine 5\\\'-monophosphate synthase protein (314-444AA)
- Antigen Synonyms:
- ODC|OMPdecase|OPRT|OPRTase|Orotate phosphoribosyl transferase and orotidine 5' decarboxylase|Orotate phosphoribosyltransferase|Orotate phosphoribosyltransferase phosphoribosyltransferase|Orotidine 5' phosphate decarboxylase|Orotidine 5''-phosphate decarboxylase|RP11-71H17.9|UMP synthase|Umps|UMPS_HUMAN|Uridine 5' monophosphate synthase|Uridine monophosphate synthetase
- ODC|OMPdecase|OPRT|OPRTase|Orotate phosphoribosyl transferase and orotidine 5' decarboxylase|Orotate phosphoribosyltransferase|Orotate phosphoribosyltransferase phosphoribosyltransferase|Orotidine 5' phosphate decarboxylase|Orotidine 5''-phosphate decarboxylase|RP11-71H17.9|UMP synthase|Umps|UMPS_HUMAN|Uridine 5' monophosphate synthase|Uridine monophosphate synthetase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11172
- P11172
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice