Anti-ZDHHC5 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ZDHHC5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-382
- 77835-381
- Antigen Symbol:
- ZDHHC5
- ZDHHC5
- Antigen Name:
- Palmitoyltransferase ZDHHC5 protein (231-500AA)
- Palmitoyltransferase ZDHHC5 protein (231-500AA)
- Antigen Synonyms:
- DKFZp586K0524|DHHC 5|DHHC-5|DHHC5|KIAA1748|Membrane associated DHHC5 zinc finger protein|MGC28179|Palmitoyltransferase ZDHHC5|Probable palmitoyltransferase ZDHHC5|ZDHC5_HUMAN|ZDHHC 5|ZDHHC5|Zinc finger DHHC domain containing 5|Zinc finger DHHC domain containing protein 5|Zinc finger DHHC domain-containing protein 5|Zinc finger DHHC type containing 5|Zinc finger protein 375|Zisp|ZNF375
- DKFZp586K0524|DHHC 5|DHHC-5|DHHC5|KIAA1748|Membrane associated DHHC5 zinc finger protein|MGC28179|Palmitoyltransferase ZDHHC5|Probable palmitoyltransferase ZDHHC5|ZDHC5_HUMAN|ZDHHC 5|ZDHHC5|Zinc finger DHHC domain containing 5|Zinc finger DHHC domain containing protein 5|Zinc finger DHHC domain-containing protein 5|Zinc finger DHHC type containing 5|Zinc finger protein 375|Zisp|ZNF375
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C0B5
- Q9C0B5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice