Anti-ADAL Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADAL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-508
- 77828-509
- Antigen Symbol:
- ADAL
- ADAL
- Antigen Name:
- Adenosine deaminase-like protein (1-267AA)
- Adenosine deaminase-like protein (1-267AA)
- Antigen Synonyms:
- ADAL|ADAL1|Adenosine deaminase-like protein|EC 3.5.4.-|Adenosine deaminase-like protein isoform 1|N6-mAMP deaminase|HsMAPDA|N6-methyl-AMP aminohydrolase
- ADAL|ADAL1|Adenosine deaminase-like protein|EC 3.5.4.-|Adenosine deaminase-like protein isoform 1|N6-mAMP deaminase|HsMAPDA|N6-methyl-AMP aminohydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6DHV7
- Q6DHV7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice