Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-SMCO1 Rabbit Recombinant Polyclonal Antibody
New Product
 
undefined
Anti-SMCO1 Rabbit Recombinant Polyclonal Antibody
New Product

 

  • Catalog No:
  • 77825-363
  • 77825-362
  • Antigen Symbol:
  • SMCO1
  • SMCO1
  • Antigen Name:
  • Single-pass membrane and coiled-coil domain-containing protein 1 protein (82-214AA)
  • Single-pass membrane and coiled-coil domain-containing protein 1 protein (82-214AA)
  • Antigen Synonyms:
  • SMCO1|C3orf43|Single-pass membrane and coiled-coil domain-containing protein 1|Single-pass membrane protein with coiled-coil domains 1
  • SMCO1|C3orf43|Single-pass membrane and coiled-coil domain-containing protein 1|Single-pass membrane protein with coiled-coil domains 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q147U7
  • Q147U7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice