Anti-TAF13 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAF13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-654
- 77822-655
- Antigen Symbol:
- TAF13
- TAF13
- Antigen Name:
- Transcription initiation factor TFIID subunit 13 protein (1-124AA)
- Transcription initiation factor TFIID subunit 13 protein (1-124AA)
- Antigen Synonyms:
- MGC22425|TAF(II)18|taf13|TAF13 RNA polymerase II|TATA box binding protein (TBP) associated factor|18kDa|TAF13_HUMAN|TAF2K|TAFII 18|TAFII-18|TAFII18|Transcription initiation factor TFIID 18 kDa subunit|Transcription initiation factor TFIID subunit 13
- MGC22425|TAF(II)18|taf13|TAF13 RNA polymerase II|TATA box binding protein (TBP) associated factor|18kDa|TAF13_HUMAN|TAF2K|TAFII 18|TAFII-18|TAFII18|Transcription initiation factor TFIID 18 kDa subunit|Transcription initiation factor TFIID subunit 13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15543
- Q15543
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice