Anti-TRIM29 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TRIM29
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77823-540
- 77823-539
- Antigen Symbol:
- TRIM29
- TRIM29
- Antigen Name:
- Tripartite motif-containing protein 29 protein (309-588AA)
- Tripartite motif-containing protein 29 protein (309-588AA)
- Antigen Synonyms:
- Ataxia telangiectasia group D associated protein|Ataxia telangiectasia group D-associated protein|ATDC|FLJ36085|TRI29_HUMAN|TRIM 29|TRIM29|Tripartite motif containing 29|Tripartite motif containing protein 29|Tripartite motif protein 29|Tripartite motif protein TRIM29|Tripartite motif-containing protein 29
- Ataxia telangiectasia group D associated protein|Ataxia telangiectasia group D-associated protein|ATDC|FLJ36085|TRI29_HUMAN|TRIM 29|TRIM29|Tripartite motif containing 29|Tripartite motif containing protein 29|Tripartite motif protein 29|Tripartite motif protein TRIM29|Tripartite motif-containing protein 29
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14134
- Q14134
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice