Anti-TMEM11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TMEM11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-829
- 77802-830
- Antigen Symbol:
- TMEM11
- TMEM11
- Antigen Name:
- Transmembrane protein 11, mitochondrial protein (1-83AA)
- Transmembrane protein 11, mitochondrial protein (1-83AA)
- Antigen Synonyms:
- C17orf35|mitochondrial|PM1|PMI|Protein PM1|Protein PMI|Putative receptor protein|tmem11|TMM11_HUMAN|Transmembrane protein 11|Transmembrane protein 11, mitochondrial
- C17orf35|mitochondrial|PM1|PMI|Protein PM1|Protein PMI|Putative receptor protein|tmem11|TMM11_HUMAN|Transmembrane protein 11|Transmembrane protein 11, mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17152
- P17152
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice