Anti-EARS2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EARS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-572
- 77827-571
- Antigen Symbol:
- EARS2
- EARS2
- Antigen Name:
- Probable glutamate--tRNA ligase, mitochondrial protein (42-166AA)
- Probable glutamate--tRNA ligase, mitochondrial protein (42-166AA)
- Antigen Synonyms:
- 3230401I01Rik|AL024049|COXPD12|EARS 2|ears2|GluRS|Glutamate--tRNA ligase|Glutamyl tRNA synthetase 2 mitochondrial|KIAA1970|mitochondrial|mKIAA1970|MSE1|Probable glutamyl tRNA synthetase|Probable glutamyl-tRNA synthetase|RGD1307904|SYEM_HUMAN
- 3230401I01Rik|AL024049|COXPD12|EARS 2|ears2|GluRS|Glutamate--tRNA ligase|Glutamyl tRNA synthetase 2 mitochondrial|KIAA1970|mitochondrial|mKIAA1970|MSE1|Probable glutamyl tRNA synthetase|Probable glutamyl-tRNA synthetase|RGD1307904|SYEM_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5JPH6
- Q5JPH6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice