Anti-PPIH Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPIH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-000
- 77795-001
- Antigen Symbol:
- PPIH
- PPIH
- Antigen Name:
- Peptidyl-prolyl cis-trans isomerase H protein (2-177AA)
- Peptidyl-prolyl cis-trans isomerase H protein (2-177AA)
- Antigen Synonyms:
- Cyclophilin H|CYP-20|CypH|Peptidyl prolyl cis trans isomerase H|Peptidyl-prolyl cis-trans isomerase H|Peptidylprolyl isomerase H|PPIase H|PPIH|PPIH_HUMAN|RGD1564921|Rotamase H|Small nuclear ribonucleoprotein particle specific cyclophilin H|Small nuclear ribonucleoprotein particle-specific cyclophilin H|SnuCyp 20|U snRNP associated cyclophilin SnuCyp 20|U-snRNP-associated cyclophilin SnuCyp-20|USA CYP|USA-CYP|USACYP
- Cyclophilin H|CYP-20|CypH|Peptidyl prolyl cis trans isomerase H|Peptidyl-prolyl cis-trans isomerase H|Peptidylprolyl isomerase H|PPIase H|PPIH|PPIH_HUMAN|RGD1564921|Rotamase H|Small nuclear ribonucleoprotein particle specific cyclophilin H|Small nuclear ribonucleoprotein particle-specific cyclophilin H|SnuCyp 20|U snRNP associated cyclophilin SnuCyp 20|U-snRNP-associated cyclophilin SnuCyp-20|USA CYP|USA-CYP|USACYP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43447
- O43447
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice