Anti-TXK Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TXK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-743
- 77811-743
- Antigen Symbol:
- TXK
- TXK
- Antigen Name:
- Tyrosine-protein kinase TXK protein (32-83AA)
- Tyrosine-protein kinase TXK protein (32-83AA)
- Antigen Synonyms:
- BTKL|EC 2.7.10.2|MGC22473|Protein-tyrosine kinase 4|PSCTK5|PTK4|Resting lymphocyte kinase|RLK|TKL|TXK|TXK tyrosine kinase|TXK_HUMAN|Tyrosine protein kinase TXK|Tyrosine-protein kinase TXK
- BTKL|EC 2.7.10.2|MGC22473|Protein-tyrosine kinase 4|PSCTK5|PTK4|Resting lymphocyte kinase|RLK|TKL|TXK|TXK tyrosine kinase|TXK_HUMAN|Tyrosine protein kinase TXK|Tyrosine-protein kinase TXK
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42681
- P42681
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice