Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anti-MED4 Rabbit Recombinant Polyclonal Antibody

Anti-MED4 Rabbit Recombinant Polyclonal Antibody

Supplier:   AFG Bioscience
Anti-MED4 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
Certificates
Anti-MED4 Rabbit Recombinant Polyclonal Antibody
Anti-MED4 Rabbit Recombinant Polyclonal Antibody

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Recommended dilution: IF:1:50 to 1:200

Type:
Antigen: MED4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications for Product Family

Specifications

  • Catalog No:
  • 77836-403
  • 77836-404
  • Antigen Symbol:
  • Activator-recruited cofactor 36 kDa component;ARC36;DRIP36;FLJ10956;HSPC126;med4;MED4_HUMAN;Mediator complex subunit 4;Mediator of RNA polymerase II transcription subunit 4;Mediator of RNA polymerase II transcription, subunit 4 homolog;OTTHUMP00000018390;OTTHUMP00000018391;OTTHUMP00000018392;RP11-90M2.2;TRAP/SMCC/PC2 subunit p36 subunit;TRAP36;VDRIP;Vitamin D3 receptor-interacting protein complex 36 kDa component
  • Activator-recruited cofactor 36 kDa component;ARC36;DRIP36;FLJ10956;HSPC126;med4;MED4_HUMAN;Mediator complex subunit 4;Mediator of RNA polymerase II transcription subunit 4;Mediator of RNA polymerase II transcription, subunit 4 homolog;OTTHUMP00000018390;OTTHUMP00000018391;OTTHUMP00000018392;RP11-90M2.2;TRAP/SMCC/PC2 subunit p36 subunit;TRAP36;VDRIP;Vitamin D3 receptor-interacting protein complex 36 kDa component
  • Antigen Name:
  • Mediator of RNA polymerase II transcription subunit 4 protein (167-270AA)
  • Mediator of RNA polymerase II transcription subunit 4 protein (167-270AA)
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NPJ6
  • Q9NPJ6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Product Family Options

Product Information

  • Size
    Packaging
    Availability
    Price
  • 100 µg
    Vial
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $333.63
    $333.63
    Each(1/Itm/Bx)
       $333.63
    Minimum order quantity is 1

    Supplier Number: A67202-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77836-403
    Antigen Symbol
    Activator-recruited cofactor 36 kDa component;ARC36;DRIP36;FLJ10956;HSPC126;med4;MED4_HUMAN;Mediator complex subunit 4;Mediator of RNA polymerase II transcription subunit 4;Mediator of RNA polymerase II transcription, subunit 4 homolog;OTTHUMP00000018390;OTTHUMP00000018391;OTTHUMP00000018392;RP11-90M2.2;TRAP/SMCC/PC2 subunit p36 subunit;TRAP36;VDRIP;Vitamin D3 receptor-interacting protein complex 36 kDa component
    Antigen Name
    Mediator of RNA polymerase II transcription subunit 4 protein (167-270AA)
    Clonality
    Recombinant Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    Q9NPJ6
    Isotype
    IgG
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Form
    Liquid
    Purification
    >95%, Protein G purified
    Storage Buffer
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
    Storage Temperature
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
    Shipping Temperature
    Shipped on blue ice
  • 50 µg
    Vial
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $227.48
    $227.48
    Each(1/Itm/Bx)
       $227.48
    Minimum order quantity is 1

    Supplier Number: A67202-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77836-404
    Antigen Symbol
    Activator-recruited cofactor 36 kDa component;ARC36;DRIP36;FLJ10956;HSPC126;med4;MED4_HUMAN;Mediator complex subunit 4;Mediator of RNA polymerase II transcription subunit 4;Mediator of RNA polymerase II transcription, subunit 4 homolog;OTTHUMP00000018390;OTTHUMP00000018391;OTTHUMP00000018392;RP11-90M2.2;TRAP/SMCC/PC2 subunit p36 subunit;TRAP36;VDRIP;Vitamin D3 receptor-interacting protein complex 36 kDa component
    Antigen Name
    Mediator of RNA polymerase II transcription subunit 4 protein (167-270AA)
    Clonality
    Recombinant Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    Q9NPJ6
    Isotype
    IgG
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Form
    Liquid
    Purification
    >95%, Protein G purified
    Storage Buffer
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
    Storage Temperature
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
    Shipping Temperature
    Shipped on blue ice
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.