Anti-PDHX Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PDHX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-601
- 77793-600
- Antigen Symbol:
- PDHX
- PDHX
- Antigen Name:
- Pyruvate dehydrogenase protein X component, mitochondrial protein (1-300AA)
- Pyruvate dehydrogenase protein X component, mitochondrial protein (1-300AA)
- Antigen Synonyms:
- Dihydrolipoamide dehydrogenase binding protein of pyruvate dehydrogenase complex|Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex|DLDBP|E3 binding protein|E3-binding protein|E3BP|Lipoyl containing pyruvate dehydrogenase complex component X|Lipoyl-containing pyruvate dehydrogenase complex component X|mitochondrial|ODPX_HUMAN|OPDX|PDHX|PDX 1|PDX1|Pro X|proX|Pyruvate dehydrogenase complex component X|Pyruvate dehydrogenase complex lipoyl containing component X|Pyruvate dehydrogenase complex|E3 binding protein subunit|Pyruvate dehydrogenase protein X component|Pyruvate dehydrogenase protein X component mitochondrial
- Dihydrolipoamide dehydrogenase binding protein of pyruvate dehydrogenase complex|Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex|DLDBP|E3 binding protein|E3-binding protein|E3BP|Lipoyl containing pyruvate dehydrogenase complex component X|Lipoyl-containing pyruvate dehydrogenase complex component X|mitochondrial|ODPX_HUMAN|OPDX|PDHX|PDX 1|PDX1|Pro X|proX|Pyruvate dehydrogenase complex component X|Pyruvate dehydrogenase complex lipoyl containing component X|Pyruvate dehydrogenase complex|E3 binding protein subunit|Pyruvate dehydrogenase protein X component|Pyruvate dehydrogenase protein X component mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00330
- O00330
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice