Anti-VPS45 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VPS45
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-944
- 77840-945
- Antigen Symbol:
- VPS45
- VPS45
- Antigen Name:
- Vacuolar protein sorting-associated protein 45 protein (101-400AA)
- Vacuolar protein sorting-associated protein 45 protein (101-400AA)
- Antigen Synonyms:
- h-VPS45|H1|H1VPS45|HGNC:14579|hlVps45|Leucocyte vacuolar protein sorting 45|OTTHUMP00000014482|Vacuolar protein sorting 45 homolog (S. cerevisiae)|Vacuolar protein sorting 45A (yeast homolog)|Vacuolar protein sorting 45A|Vacuolar protein sorting 45B (yeast)|Vacuolar protein sorting-associated protein 45|Vps45|VPS45_HUMAN|VPS45A|VPS45B|VPS54A|VSP45|VSP45A
- h-VPS45|H1|H1VPS45|HGNC:14579|hlVps45|Leucocyte vacuolar protein sorting 45|OTTHUMP00000014482|Vacuolar protein sorting 45 homolog (S. cerevisiae)|Vacuolar protein sorting 45A (yeast homolog)|Vacuolar protein sorting 45A|Vacuolar protein sorting 45B (yeast)|Vacuolar protein sorting-associated protein 45|Vps45|VPS45_HUMAN|VPS45A|VPS45B|VPS54A|VSP45|VSP45A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NRW7
- Q9NRW7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice