Anti-PRKAG3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77841-357
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:5\\\'-AMP-activated protein kinase subunit gamma-3 protein (1-210AA)
- Antigen Symbol:PRKAG3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9UGI9
- Antigen Synonyms:5 AMP activated protein kinase subunit gamma 3|5''-AMP-activated protein kinase subunit gamma-3|AAKG3_HUMAN|AMPK gamma 3 chain|AMPK gamma3|AMPK subunit gamma-3|AMPKG3|PRKAG3|Protein kinase AMP activated gamma 3 non catalytic subunit
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: PRKAG3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat