Anti-POLG2 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: POLG2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-396
- 77841-397
- Antigen Symbol:
- POLG2
- POLG2
- Antigen Name:
- DNA polymerase subunit gamma-2, mitochondrial protein (1-485AA)
- DNA polymerase subunit gamma-2, mitochondrial protein (1-485AA)
- Antigen Synonyms:
- DNA directed DNA polymerase gamma 2|DNA polymerase gamma accessory 55 kDa subunit|DNA polymerase subunit gamma 2|mitochondrial|DNA polymerase subunit gamma-2|DPOG2_HUMAN|HP55|Mitochondrial DNA polymerase accessory subunit|Mitochondrial DNA polymerase subunit gamma 2|Mitochondrial polymerase gamma 2|MtPolB|OTTMUSP00000003758|p55|PEOA4|POLB|PolG beta|PolG-beta|POLG2|POLGB|Polymerase (DNA directed)|gamma 2|accessory subunit|Polymerase|RP23-247J12.3
- DNA directed DNA polymerase gamma 2|DNA polymerase gamma accessory 55 kDa subunit|DNA polymerase subunit gamma 2|mitochondrial|DNA polymerase subunit gamma-2|DPOG2_HUMAN|HP55|Mitochondrial DNA polymerase accessory subunit|Mitochondrial DNA polymerase subunit gamma 2|Mitochondrial polymerase gamma 2|MtPolB|OTTMUSP00000003758|p55|PEOA4|POLB|PolG beta|PolG-beta|POLG2|POLGB|Polymerase (DNA directed)|gamma 2|accessory subunit|Polymerase|RP23-247J12.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHN1
- Q9UHN1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice