Anti-DUSP13 Rabbit Recombinant Polyclonal Antibody
Catalog # 77829-108
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Dual specificity protein phosphatase 13 isoform A protein (1-198AA)
- Antigen Symbol:DUSP13
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q6B8I1
- Antigen Synonyms:DUSP13|BEDP|DUSP13A|MDSPDual specificity protein phosphatase 13 isoform A|EC 3.1.3.16|EC 3.1.3.48|Branching-enzyme interacting DSP|Muscle-restricted DSP|MDSP
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: DUSP13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human