Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FHL2 Rabbit Polyclonal Antibody
Anti-FHL2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FHL2 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77826-243
  • 77826-244
  • Antigen Symbol:
  • FHL2
  • FHL2
  • Antigen Name:
  • Fusion protein of FHL2
  • Fusion protein of FHL2
  • Antigen Synonyms:
  • AAG 11|AAG11|Aging associated gene 11|Down regulated in rhabdomyosarcoma LIM protein|Downregulated in rhabdomyosarcoma LIM protein|DRAL|FHL 2|FHL-2|Fhl2|FHL2 protein|FHL2_HUMAN|Four and a half LIM domain protein 2|Four and a half LIM domains 2|Four and a half LIM domains protein 2|KIAA0990|LIM domain protein DRAL|Skeletal muscle LIM protein 3|Skeletal muscle LIM-protein 3|SLIM 3|SLIM-3|SLIM3
  • AAG 11|AAG11|Aging associated gene 11|Down regulated in rhabdomyosarcoma LIM protein|Downregulated in rhabdomyosarcoma LIM protein|DRAL|FHL 2|FHL-2|Fhl2|FHL2 protein|FHL2_HUMAN|Four and a half LIM domain protein 2|Four and a half LIM domains 2|Four and a half LIM domains protein 2|KIAA0990|LIM domain protein DRAL|Skeletal muscle LIM protein 3|Skeletal muscle LIM-protein 3|SLIM 3|SLIM-3|SLIM3
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Mouse, Rat
  • Human, Mouse, Rat
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q14192
  • Q14192
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications