Anti-TRIM9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TRIM9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-436
- 77840-437
- Antigen Symbol:
- TRIM9
- TRIM9
- Antigen Name:
- E3 ubiquitin-protein ligase TRIM9 protein (1-550AA)
- E3 ubiquitin-protein ligase TRIM9 protein (1-550AA)
- Antigen Synonyms:
- TRIM9|KIAA0282|RNF91|E3 ubiquitin-protein ligase TRIM9|EC 2.3.2.27|RING finger protein 91|RING-type E3 ubiquitin transferase TRIM9|Tripartite motif-containing protein 9
- TRIM9|KIAA0282|RNF91|E3 ubiquitin-protein ligase TRIM9|EC 2.3.2.27|RING finger protein 91|RING-type E3 ubiquitin transferase TRIM9|Tripartite motif-containing protein 9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C026
- Q9C026
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice