Anti-MRM3 Rabbit Polyclonal Antibody
Catalog # 77797-598
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:RNMTL1
- Antigen Symbol:MRM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9HC36
- Antigen Synonyms:16S rRNA (guanosine(1370)-2'-O)-methyltransferase|16S rRNA [Gm1370] 2'-O-methyltransferase|4833420N02Rik|AI255153|FLJ10581|HC90|MRM3|MRM3_HUMAN|Putative RNA methyltransferase|RMTL1|RNA methyltransferase like 1|RNA methyltransferase like protein 1|RNA methyltransferase-like protein 1|RP23-147P4.7|rRNA methyltransferase 3, mitochondrial
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MRM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat