Anti-ZNF699 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: ZNF699
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-805
- 77805-804
- Antigen Symbol:
- ZNF699
- ZNF699
- Antigen Name:
- Zinc finger protein 699 protein (19-195AA)
- Zinc finger protein 699 protein (19-195AA)
- Antigen Synonyms:
- FLJ38144|hang|Hangover homolog|hangover|Drosophila|homolog of|MGC129880|MGC129881|Zinc finger protein 699|ZN699_HUMAN
- FLJ38144|hang|Hangover homolog|hangover|Drosophila|homolog of|MGC129880|MGC129881|Zinc finger protein 699|ZN699_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q32M78
- Q32M78
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice