Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ZCCHC17 Rabbit Recombinant Polyclonal Antibody
Anti-ZCCHC17 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ZCCHC17 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77839-701
  • 77839-700
  • Antigen Symbol:
  • ZCCHC17
  • ZCCHC17
  • Antigen Name:
  • Nucleolar protein of 40 kDa protein (1-241AA)
  • Nucleolar protein of 40 kDa protein (1-241AA)
  • Antigen Synonyms:
  • 2810055E05Rik|HSPC251|LDC4|NO40_HUMAN|Nucleolar protein 40|Nucleolar protein of 40 kDa|OTTHUMP00000003890|OTTHUMP00000003891|Pnn-interacting nucleolar protein|pNO40|pS1D|PS1D protein|Putative S1 RNA binding domain protein|Putative S1 RNA-binding domain protein|RGD1565267|ZCCHC17|Zinc finger CCHC domain-containing protein 17|Zinc finger|CCHC domain containing 17
  • 2810055E05Rik|HSPC251|LDC4|NO40_HUMAN|Nucleolar protein 40|Nucleolar protein of 40 kDa|OTTHUMP00000003890|OTTHUMP00000003891|Pnn-interacting nucleolar protein|pNO40|pS1D|PS1D protein|Putative S1 RNA binding domain protein|Putative S1 RNA-binding domain protein|RGD1565267|ZCCHC17|Zinc finger CCHC domain-containing protein 17|Zinc finger|CCHC domain containing 17
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9NP64
  • Q9NP64
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications