Anti-ZNF689 Rabbit Recombinant Polyclonal Antibody
77805-788
New Product
- Antigen Name:Zinc finger protein 689 protein (1-300AA)
- Antigen Symbol:ZNF689
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96CS4
- Antigen Synonyms:ZNF689|Zinc finger protein 689
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: ZNF689
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human