Anti-PURA Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PURA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-217
- 77796-216
- Antigen Symbol:
- PURA
- PURA
- Antigen Name:
- Transcriptional activator protein Pur-alpha protein (50-296AA)
- Transcriptional activator protein Pur-alpha protein (50-296AA)
- Antigen Synonyms:
- PUR ALPHA|PUR1|purA|PURA_HUMAN|PURALPHA|Purine rich element binding protein A|Purine rich single stranded DNA binding protein alpha|Purine-rich single-stranded DNA-binding protein alpha|Transcriptional activator protein PUR alpha|Transcriptional activator protein Pur-alpha
- PUR ALPHA|PUR1|purA|PURA_HUMAN|PURALPHA|Purine rich element binding protein A|Purine rich single stranded DNA binding protein alpha|Purine-rich single-stranded DNA-binding protein alpha|Transcriptional activator protein PUR alpha|Transcriptional activator protein Pur-alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00577
- Q00577
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice