Anti-PCID2 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: PCID2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-582
- 77827-581
- Antigen Symbol:
- PCID2
- PCID2
- Antigen Name:
- PCI domain-containing protein 2 protein (2-250AA)
- PCI domain-containing protein 2 protein (2-250AA)
- Antigen Synonyms:
- CSN12 like protein|CSN12-like protein|DKFZp686C20226|F10|FLJ11305|FLJ99362|HT004|MGC16774|PCI domain containing 2|PCI domain containing protein 2|PCI domain-containing protein 2|PCID2|PCID2_HUMAN|RP11 - 98F14.6
- CSN12 like protein|CSN12-like protein|DKFZp686C20226|F10|FLJ11305|FLJ99362|HT004|MGC16774|PCI domain containing 2|PCI domain containing protein 2|PCI domain-containing protein 2|PCID2|PCID2_HUMAN|RP11 - 98F14.6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5JVF3
- Q5JVF3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice