Anti-ACSBG1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACSBG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-046
- 77833-045
- Antigen Symbol:
- ACSBG1
- ACSBG1
- Antigen Name:
- Long-chain-fatty-acid--CoA ligase ACSBG1 protein (351-600AA)
- Long-chain-fatty-acid--CoA ligase ACSBG1 protein (351-600AA)
- Antigen Synonyms:
- ACBG1_HUMAN|ACSBG 1||Acyl CoA synthetase bubblegum family member 1|Acyl-CoA synthetase bubblegum family member 1|BG|BG1|BGM|Bubblegum|FLJ30320|GR LACS|GRLACS|hBG1|hsBG|hsBGM|KIAA0631|Lipidosin|Lipodisin|Long chain fatty acid CoA ligase |Long-chain-fatty-acid--CoA ligase |LPD|MGC14352|Ortholog of rodent gonadotropin regulated long chain acyl CoA synthetase|Very long chain acyl CoA synthetase
- ACBG1_HUMAN|ACSBG 1||Acyl CoA synthetase bubblegum family member 1|Acyl-CoA synthetase bubblegum family member 1|BG|BG1|BGM|Bubblegum|FLJ30320|GR LACS|GRLACS|hBG1|hsBG|hsBGM|KIAA0631|Lipidosin|Lipodisin|Long chain fatty acid CoA ligase |Long-chain-fatty-acid--CoA ligase |LPD|MGC14352|Ortholog of rodent gonadotropin regulated long chain acyl CoA synthetase|Very long chain acyl CoA synthetase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GR2
- Q96GR2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice