Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SMC1B Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-SMC1B Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SMC1B Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77833-447
  • 77833-448
  • Antigen Symbol:
  • SMC1B
  • SMC1B
  • Antigen Name:
  • Structural maintenance of chromosomes protein 1B protein (954-1060AA)
  • Structural maintenance of chromosomes protein 1B protein (954-1060AA)
  • Antigen Synonyms:
  • bK268H5|SMC protein 1B|SMC-1-beta|SMC-1B|SMC1 beta|SMC1B|SMC1B_HUMAN|SMC1beta|SMC1beta protein|Smc1l2|Structural maintenance of chromosomes 1-like 2|Structural maintenance of chromosomes 1B|Structural maintenance of chromosomes protein 1B
  • bK268H5|SMC protein 1B|SMC-1-beta|SMC-1B|SMC1 beta|SMC1B|SMC1B_HUMAN|SMC1beta|SMC1beta protein|Smc1l2|Structural maintenance of chromosomes 1-like 2|Structural maintenance of chromosomes 1B|Structural maintenance of chromosomes protein 1B
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8NDV3
  • Q8NDV3
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together