Anti-RAB31 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RAB31
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-436
- 77822-435
- Antigen Symbol:
- RAB31
- RAB31
- Antigen Name:
- Ras-related protein Rab-31 protein (1-194AA)
- Ras-related protein Rab-31 protein (1-194AA)
- Antigen Synonyms:
- Rab 22B|RAB 31|Rab22B|Rab31|RAB31 member RAS oncogene family|RAB31_HUMAN|Ras related protein Rab 31|Ras related protein Rab31|Ras-related protein Rab-22B|Ras-related protein Rab-31
- Rab 22B|RAB 31|Rab22B|Rab31|RAB31 member RAS oncogene family|RAB31_HUMAN|Ras related protein Rab 31|Ras related protein Rab31|Ras-related protein Rab-22B|Ras-related protein Rab-31
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13636
- Q13636
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice