Anti-LAMTOR1 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LAMTOR1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-822
- 77830-823
- Antigen Symbol:
- LAMTOR1
- LAMTOR1
- Antigen Name:
- Ragulator complex protein LAMTOR1 protein (2-161AA)
- Ragulator complex protein LAMTOR1 protein (2-161AA)
- Antigen Synonyms:
- C11orf59|LAMTOR1|Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1|Lipid raft adaptor protein p18|LTOR1_HUMAN|p18|p27Kip1-releasing factor from RhoA|p27RF-Rho|PDRO|PP7157|Protein associated with DRMs and endosomes|Ragulator complex protein LAMTOR1|ragulator complex protein PDRO|Ragulator1|RhoA activator C11orf59
- C11orf59|LAMTOR1|Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1|Lipid raft adaptor protein p18|LTOR1_HUMAN|p18|p27Kip1-releasing factor from RhoA|p27RF-Rho|PDRO|PP7157|Protein associated with DRMs and endosomes|Ragulator complex protein LAMTOR1|ragulator complex protein PDRO|Ragulator1|RhoA activator C11orf59
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6IAA8
- Q6IAA8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice