Anti-PPP2R5E Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPP2R5E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-765
- 77823-764
- Antigen Symbol:
- PPP2R5E
- PPP2R5E
- Antigen Name:
- Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform protein (2-149AA)
- Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform protein (2-149AA)
- Antigen Synonyms:
- 2A5E_HUMAN|Epsilon isoform of regulatory subunit B56 protein phosphatase 2A|PP2A B subunit B' epsilon|PP2A B subunit B' epsilon isoform|PP2A B subunit B56 epsilon|PP2A B subunit B56 epsilon isoform|PP2A B subunit isoform B''-epsilon|PP2A B subunit isoform B'-epsilon|PP2A B subunit isoform B56-epsilon|PP2A B subunit isoform PR61-epsilon|PP2A B subunit isoform R5-epsilon|PP2A B subunit PR61 epsilon|PP2A B subunit PR61 epsilon isoform|PP2A B subunit R5 epsilon|PP2A B subunit R5 epsilon isoform|PPP2R5E|Protein phosphatase 2 regulatory subunit B (B56) epsilon isoform|Protein phosphatase 2 regulatory subunit B' epsilon|Protein phosphatase 2 regulatory subunit B' epsilon isoform|Regulatory subunit B of protein phosphatase 2 epsilon isoform|Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit epsilon|Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform
- 2A5E_HUMAN|Epsilon isoform of regulatory subunit B56 protein phosphatase 2A|PP2A B subunit B' epsilon|PP2A B subunit B' epsilon isoform|PP2A B subunit B56 epsilon|PP2A B subunit B56 epsilon isoform|PP2A B subunit isoform B''-epsilon|PP2A B subunit isoform B'-epsilon|PP2A B subunit isoform B56-epsilon|PP2A B subunit isoform PR61-epsilon|PP2A B subunit isoform R5-epsilon|PP2A B subunit PR61 epsilon|PP2A B subunit PR61 epsilon isoform|PP2A B subunit R5 epsilon|PP2A B subunit R5 epsilon isoform|PPP2R5E|Protein phosphatase 2 regulatory subunit B (B56) epsilon isoform|Protein phosphatase 2 regulatory subunit B' epsilon|Protein phosphatase 2 regulatory subunit B' epsilon isoform|Regulatory subunit B of protein phosphatase 2 epsilon isoform|Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit epsilon|Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit epsilon isoform
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16537
- Q16537
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice