Anti-MAT2B Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAT2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-170
- 77841-171
- Antigen Symbol:
- MAT2B
- MAT2B
- Antigen Name:
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Antigen Synonyms:
- 1110064C04Rik| 2410018D16Rik| AI182287| AU022853| Beta regulatory subunit of methionine adenosyltransferase| dTDP 4 keto 6 deoxy D glucose 4 reductase| DTDP-4-keto-6-deoxy-D-glucose 4-reductase| MAT II| MAT II beta| Mat2b| MAT2B_HUMAN| MATIIbeta| Methionine adenosyltransferase 2 beta subunit| Methionine adenosyltransferase 2 subunit beta| Methionine adenosyltransferase II beta| MGC12237| MSTP045| Nbla02999| OTTMUSP00000005600| Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase| Putative protein product of Nbla02999| RP23-382C18.2| SDR23E1| Short chain dehydrogenase/reductase family 23E member 1| TGR
- 1110064C04Rik| 2410018D16Rik| AI182287| AU022853| Beta regulatory subunit of methionine adenosyltransferase| dTDP 4 keto 6 deoxy D glucose 4 reductase| DTDP-4-keto-6-deoxy-D-glucose 4-reductase| MAT II| MAT II beta| Mat2b| MAT2B_HUMAN| MATIIbeta| Methionine adenosyltransferase 2 beta subunit| Methionine adenosyltransferase 2 subunit beta| Methionine adenosyltransferase II beta| MGC12237| MSTP045| Nbla02999| OTTMUSP00000005600| Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase| Putative protein product of Nbla02999| RP23-382C18.2| SDR23E1| Short chain dehydrogenase/reductase family 23E member 1| TGR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZL9
- Q9NZL9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice