Anti-SLC13A3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: SLC13A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-513
- 77818-512
- Antigen Symbol:
- SLC13A3
- SLC13A3
- Antigen Name:
- Synthetic peptide of SLC13A3
- Synthetic peptide of SLC13A3
- Antigen Synonyms:
- dicarboxylate cotransporter 3|H sapiens mRNA for hepatocyte nuclear factor 4 gamma|hNaDC3|Na|Na(+)/dicarboxylate cotransporter 3|NaDC 3|NaDC-3|NADC3|S13A3_HUMAN|SDCT 2|SDCT2|SLC13A3|Sodium dependent high affinity dicarboxylate transporter 2|Sodium dependent high affinity dicarboxylate transporter 3|Sodium-dependent high-affinity dicarboxylate transporter 2|Solute carrier family 13 (sodium dependent dicarboxylate transporter)|member 3|Solute carrier family 13 member 3
- dicarboxylate cotransporter 3|H sapiens mRNA for hepatocyte nuclear factor 4 gamma|hNaDC3|Na|Na(+)/dicarboxylate cotransporter 3|NaDC 3|NaDC-3|NADC3|S13A3_HUMAN|SDCT 2|SDCT2|SLC13A3|Sodium dependent high affinity dicarboxylate transporter 2|Sodium dependent high affinity dicarboxylate transporter 3|Sodium-dependent high-affinity dicarboxylate transporter 2|Solute carrier family 13 (sodium dependent dicarboxylate transporter)|member 3|Solute carrier family 13 member 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WWT9
- Q8WWT9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice