Anti-USP17L1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: USP17L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-808
- 77830-809
- Antigen Symbol:
- USP17L1
- USP17L1
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 17-like protein 1 protein (19-235AA)
- Ubiquitin carboxyl-terminal hydrolase 17-like protein 1 protein (19-235AA)
- Antigen Synonyms:
- USP17L1|USP17L|USP17L1P|Ubiquitin carboxyl-terminal hydrolase 17-like protein 1|EC 3.4.19.12|Deubiquitinating enzyme 17-like protein 1|Ubiquitin thioesterase 17-like protein 1|Ubiquitin-specific-processing protease 17-like protein 1
- USP17L1|USP17L|USP17L1P|Ubiquitin carboxyl-terminal hydrolase 17-like protein 1|EC 3.4.19.12|Deubiquitinating enzyme 17-like protein 1|Ubiquitin thioesterase 17-like protein 1|Ubiquitin-specific-processing protease 17-like protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7RTZ2
- Q7RTZ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice