Anti-KRTAP13-4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
About this item
Type:
Antigen: KRTAP13-4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: KRTAP13-4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications for Product Family
Specifications
- Catalog No:
- 77826-296
- 77826-297
- Antigen Symbol:
- KRTAP13-4
- KRTAP13-4
- Antigen Name:
- Keratin-associated protein 13-4 protein (1-160AA)
- Keratin-associated protein 13-4 protein (1-160AA)
- Antigen Synonyms:
- KRTAP13-4|KAP13.4Keratin-associated protein 13-4
- KRTAP13-4|KAP13.4Keratin-associated protein 13-4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3LI77
- Q3LI77
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Product Family Options
Product Information
- SizePackagingAvailabilityPrice
- Supplier Number: A56447-100UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77826-296Antigen SymbolKRTAP13-4Antigen NameKeratin-associated protein 13-4 protein (1-160AA)Antigen SynonymsKRTAP13-4|KAP13.4Keratin-associated protein 13-4ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ3LI77IsotypeIgGELISAYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice - Supplier Number: A56447-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.77826-297Antigen SymbolKRTAP13-4Antigen NameKeratin-associated protein 13-4 protein (1-160AA)Antigen SynonymsKRTAP13-4|KAP13.4Keratin-associated protein 13-4ClonalityRecombinant PolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene IDQ3LI77IsotypeIgGELISAYesFormLiquidPurification>95%, Protein G purifiedStorage Buffer0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4Storage TemperatureUpon receipt, store at −20 or −80 °C. Avoid repeated freeze.Shipping TemperatureShipped on blue ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



