Anti-PAK1/PAK2/PAK3 Rabbit Polyclonal Antibody
77820-965
New Product
- Antigen Name:STE20 homolog yeast
- Antigen Symbol:PAK1/PAK2/PAK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13153
- Antigen Synonyms:ADRB2|Alpha PAK|Alpha-PAK|MGC130000|MGC130001|p21 activated kinase 1|p21 protein (Cdc42/Rac) activated kinase 1|p21-activated kinase 1|p21/Cdc42/Rac1 activated kinase 1 (yeast Ste20 related)|p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast)|p65 PAK|p65-PAK|P68-PAK|PAK alpha|PAK-1|Pak1|PAK1_HUMAN|Paka|PAKalpha|Protein kinase MUK2|Rac/p21-activated kinase|Serine/threonine-protein kinase PAK 1|STE20 homolog yeast
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200, IF: 1:100 to 1:200
Type:
Antigen: PAK1/PAK2/PAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat