Anti-SPZ1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SPZ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77842-945
- 77842-944
- Antigen Symbol:
- SPZ1
- SPZ1
- Antigen Name:
- Spermatogenic leucine zipper protein 1 protein (181-430AA)
- Spermatogenic leucine zipper protein 1 protein (181-430AA)
- Antigen Synonyms:
- SPZ1|TSP1|Spermatogenic leucine zipper protein 1|Testis-specific protein 1|Testis-specific protein NYD-TSP1
- SPZ1|TSP1|Spermatogenic leucine zipper protein 1|Testis-specific protein 1|Testis-specific protein NYD-TSP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXG8
- Q9BXG8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice