Anti-KLHL13 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KLHL13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-656
- 77836-655
- Antigen Symbol:
- KLHL13
- KLHL13
- Antigen Name:
- Kelch-like protein 13 protein (1-260AA)
- Kelch-like protein 13 protein (1-260AA)
- Antigen Synonyms:
- BKLHD2|BTB and kelch domain containing 2|BTB and kelch domain containing 2 KIAA1309|BTB and kelch domain containing protein 2|BTB and kelch domain-containing protein 2|FLJ10262|Kelch like 13 (Drosophila)|Kelch like protein 13|Kelch-like protein 13|KIAA1309|KLH13_HUMAN||MGC74791
- BKLHD2|BTB and kelch domain containing 2|BTB and kelch domain containing 2 KIAA1309|BTB and kelch domain containing protein 2|BTB and kelch domain-containing protein 2|FLJ10262|Kelch like 13 (Drosophila)|Kelch like protein 13|Kelch-like protein 13|KIAA1309|KLH13_HUMAN||MGC74791
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2N7
- Q9P2N7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice