Anti-ZADH2 Rabbit Polyclonal Antibody
100 µl, Vial
Catalog # 77804-874
Supplier: AFG Bioscience
Supplier #: A38635-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type:
Antigen: ZADH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Specifications
- Size:100 µl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77804-874
- Reactivity:Human|Mouse|Rat
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:ZADH2
- Gene ID:Q8N4Q0
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:ZADH2|PTGR3|Prostaglandin reductase 3|PRG-3|EC 1.3.1.48|Zinc-binding alcohol dehydrogenase domain-containing protein 2
- Antigen Name:Zinc-binding alcohol dehydrogenase domain-containing protein 2
- ELISA:Yes
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



