Anti-CHRD Rabbit Polyclonal Antibody
Catalog # 77839-292
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthetic peptide from protein at AA range: 905-954
- Antigen Symbol:CHRD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H2X0
- Antigen Synonyms:CHRD|UNQ217/PRO243Chordin
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 300, ELISA: 1:10000 to 20000
Type:
Antigen: CHRD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat